Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032558.2 | NP_115947.2 | 12 | 2893 | ||
XM_006717305.3 | XP_006717368.1 | 11 | 2770 | |||
XM_011519114.2 | XP_011517416.1 | 12 | 2584 | |||
XM_017015217.1 | XP_016870706.1 | 12 | 2694 | |||
XM_017015218.1 | XP_016870707.1 | 12 | 2621 | |||
XM_017015219.1 | XP_016870708.1 | 11 | 2431 | |||
XM_017015220.1 | XP_016870709.1 | 11 | 2445 | |||
XM_017015221.1 | XP_016870710.1 | 11 | 2486 | |||
GenBank mRNA | AF255650.1 | - | 6 | 1868 | ||
AK027659.1 | - | 8 | 2197 | |||
AL832415.1 | - | 5 | 1760 | |||
BC106062.1 | - | 12 | 2630 | 6162855 | ||
BX648846.1 | - | 9 | 2304 |