Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278740.1 | NP_001265669.1 | 2 | 592 | ||
NM_001278741.1 | NP_001265670.1 | 2 | 362 | |||
NM_001278742.1 | NP_001265671.1 | 2 | 362 | |||
NM_001278743.1 | NP_001265672.1 | 2 | 362 | |||
NM_003270.3 | NP_003261.1 | 2 | 300 | |||
XM_011531018.1 | XP_011529320.1 | 3 | 102 | |||
GenBank mRNA | AB097016.1 | - | 2 | 196 | ||
AB209876.1 | - | 2 | 259 | |||
AF043906.1 | - | 2 | 204 | |||
AF053453.1 | - | 2 | 156 | |||
AF133426.1 | - | 2 | 163 | |||
AK075072.1 | - | 2 | 249 | |||
AK309581.1 | - | 2 | 575 | |||
AK315773.1 | - | 2 | 281 | |||
AY358825.1 | - | 2 | 177 | |||
BC012389.1 | - | 2 | 240 | 3857537 | ||
BT006977.1 | - | 2 | 137 | |||
CN274931.1 | - | 2 | 82 | |||
CR457115.1 | - | 2 | 137 | |||
CX760536.1 | - | 2 | 300 | |||
CX868833.1 | - | 2 | 71 | |||
DA038474.1 | - | 2 | 99 | |||
U84895.1 | - | 2 | 204 |