Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288622.1 | NP_001275551.1 | 13 | 7195 | ||
NM_001288623.1 | NP_001275552.1 | 13 | 7140 | |||
NM_138468.5 | NP_612477.3 | 14 | 7322 | |||
GenBank mRNA | AK124756.1 | - | 1 | 2197 |
Gene Symbol: PIGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042616.2 | NP_001036081.1 | 1, 2 | 304 | ||
GenBank mRNA | AB206972.1 | - | 1, 2 | 276 | ||
BC007876.2 | - | 1, 2 | 276 | 3947944 | ||
BI548853.1 | - | 1, 2 | 309 |
Gene Symbol: PYURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032906.4 | NP_116295.1 | 1, 2 | 304 | ||
GenBank mRNA | AK312082.1 | - | 1, 2 | 279 | ||
BG706896.1 | - | 1, 2 | 309 | |||
CV812938.1 | - | 1, 2 | 240 |