Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277233.1 | NP_001264162.1 | 3 | 434 | ||
NM_003273.3 | NP_003264.2 | 3 | 434 | |||
GenBank mRNA | AF023676.1 | - | 1 | 928 | ||
AF096304.1 | - | 3 | 398 | |||
AK290935.1 | - | 3 | 412 | |||
BC009052.1 | - | 3 | 400 | 3874367 | ||
BC012857.1 | - | 3 | 362 | 4214662 | ||
BC038353.1 | - | 3 | 404 | 3861907 | ||
BG771219.1 | - | 3 | 420 |