Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166116.1 | NP_001159588.1 | 2 | 227 | ||
NM_003275.3 | NP_003266.1 | 2 | 304 | |||
GenBank mRNA | AK314533.1 | - | 2 | 227 | ||
BC002660.2 | - | 2 | 304 | 3610014 | ||
DA015088.1 | - | 2 | 277 | |||
M77016.1 | - | 1 | 125 |