Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033387.3 | NP_203745.2 | 2 | 2123 | ||
XM_011518567.1 | XP_011516869.1 | 3 | 1666 | |||
XM_011518568.2 | XP_011516870.1 | 2 | 2369 | |||
GenBank mRNA | AK024434.1 | - | 3 | 2745 | ||
BC029924.2 | - | 2 | 1537 | |||
BC049388.2 | - | 2 | 2123 | 6050655 |