Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163321.2 | NP_001156793.2 | 11 | 3443 | ||
NM_001163322.2 | NP_001156794.1 | 11 | 3384 | |||
NM_001163323.2 | NP_001156795.1 | 10 | 3873 | |||
NM_001271835.1 | NP_001258764.1 | 10 | 3873 | |||
NM_001271836.1 | NP_001258765.1 | 10 | 4225 | |||
NM_033626.3 | NP_296375.1 | 10 | 3814 | |||
GenBank mRNA | AK056290.1 | - | 1 | 2410 | ||
AK125424.1 | - | 3408 | ||||
BC008769.2 | - | 10 | 3795 | 3139519 |