Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052864.2 | NP_443096.1 | 2 | 885 | ||
GenBank mRNA | AB097011.1 | - | 2 | 887 | ||
AB097038.1 | - | 2 | 887 | |||
AK292900.1 | - | 2 | 882 | |||
BC008294.1 | - | 2 | 870 | |||
BC014259.2 | - | 2 | 865 | 4763971 |