Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024380.1 | NP_001019551.1 | 4 | 1171 | ||
NM_001024381.1 | NP_001019552.1 | 4 | 1070 | |||
NM_031940.3 | NP_114146.3 | 4 | 1377 | |||
NM_078473.2 | NP_510882.1 | 4 | 988 | |||
XM_005273657.3 | XP_005273714.1 | 4 | 1271 | |||
XM_006716408.3 | XP_006716471.1 | 4 | 1153 | |||
XM_011544667.2 | XP_011542969.1 | 4 | 1513 | |||
GenBank mRNA | AF353991.1 | - | 4 | 958 | ||
AK075027.1 | - | 4 | 1349 | |||
AK075155.1 | - | 4 | 1372 | |||
AL834224.1 | - | 4 | 1031 | |||
BC004878.2 | - | 4 | 1340 | 3688720 |