Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198805.1 | NP_001185734.1 | 3 | 855 | ||
NM_001198806.1 | NP_001185735.1 | 3 | 862 | |||
NM_001198807.1 | NP_001185736.1 | 4 | 963 | |||
NM_080655.2 | NP_542386.1 | 3 | 894 | |||
GenBank mRNA | AK092292.1 | - | 3 | 1580 | ||
AK314924.1 | - | 3 | 754 | |||
AY598327.1 | - | 2 | 680 | |||
BC008993.1 | - | 3 | 747 | 4213591 |