Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100Z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130772.3 | NP_570128.2 | 4 | 493 | ||
XM_011543239.2 | XP_011541541.1 | 4 | 541 | |||
XM_011543240.2 | XP_011541542.1 | 4 | 541 | |||
XM_011543241.2 | XP_011541543.1 | 4 | 541 | |||
XM_011543244.2 | XP_011541546.1 | 4 | 508 | |||
XM_011543245.2 | XP_011541547.1 | 3 | 1278 | |||
XM_017009171.1 | XP_016864660.1 | 4 | 733 | |||
XM_017009172.1 | XP_016864661.1 | 4 | 744 | |||
XM_017009173.1 | XP_016864662.1 | 3 | 422 | |||
GenBank mRNA | AF437876.1 | - | 2 | 233 | ||
BC022320.1 | - | 4 | 494 | 4696667 | ||
BX281742.1 | - | 4 | 395 |