Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM104B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166699.1 | NP_001160171.1 | 4 | 746 | ||
NM_138362.3 | NP_612371.2 | 4 | 743 | |||
NR_030722.1 | 4 | 750 | ||||
XM_017029951.1 | XP_016885440.1 | 3 | 3072 | |||
XM_017029952.1 | XP_016885441.1 | 3 | 3285 | |||
XM_017029953.1 | XP_016885442.1 | 4 | 840 | |||
GenBank mRNA | AK000441.1 | - | 4 | 727 | ||
BC000919.2 | - | 4 | 721 | 3449838 | ||
BC017571.1 | - | 4 | 731 | 4105917 | ||
BI850888.1 | - | 1 | 396 |