Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159651.1 | NP_001153123.1 | 3 | 953 | ||
NM_138369.2 | NP_612378.1 | 4 | 1150 | |||
XM_017010017.1 | XP_016865506.1 | 2 | 1242 | |||
GenBank mRNA | AK301982.1 | - | 4 | 1138 | ||
BC003114.1 | - | 3 | 878 | 3537973 | ||
BC007436.2 | - | 4 | 1140 | 2961368 |