Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138386.2 | NP_612395.2 | 8 | 1301 | ||
XM_011532410.2 | XP_011530712.1 | 8 | 4768 | |||
XM_011532411.2 | XP_011530713.1 | 9 | 1375 | |||
GenBank mRNA | BC008207.1 | - | 8 | 1301 | 3447276 |