Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 12 | 3958 | ||
XM_005263115.3 | XP_005263172.1 | 12 | 4013 | |||
XM_011532091.2 | XP_011530393.1 | 11 | 3907 | |||
XM_011532092.2 | XP_011530394.1 | 11 | 3838 | |||
XM_011532093.2 | XP_011530395.1 | 11 | 3784 | |||
XM_011532094.2 | XP_011530396.1 | 10 | 3700 | |||
XM_011532096.2 | XP_011530398.1 | 12 | 3958 | |||
XM_011532097.2 | XP_011530399.1 | 5 | 1060 | |||
XM_011532098.2 | XP_011530400.1 | 3 | 734 | |||
XM_011532099.2 | XP_011530401.1 | 2 | 693 | |||
XM_017008369.1 | XP_016863858.1 | 11 | 3953 | |||
XM_017008370.1 | XP_016863859.1 | 10 | 3733 | |||
XM_017008371.1 | XP_016863860.1 | 9 | 3613 | |||
GenBank mRNA | AK126054.1 | - | 1 | 342 | ||
BX647594.1 | - | 1 | 441 | |||
CN297341.1 | - | 1 | 557 |