Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 15 | 4354 | ||
XM_005263115.3 | XP_005263172.1 | 15 | 4409 | |||
XM_011532091.2 | XP_011530393.1 | 14 | 4303 | |||
XM_011532092.2 | XP_011530394.1 | 14 | 4234 | |||
XM_011532093.2 | XP_011530395.1 | 14 | 4180 | |||
XM_011532094.2 | XP_011530396.1 | 13 | 4096 | |||
XM_011532096.2 | XP_011530398.1 | 15 | 4354 | |||
XM_011532097.2 | XP_011530399.1 | 8 | 1456 | |||
XM_011532098.2 | XP_011530400.1 | 6 | 1130 | |||
XM_011532099.2 | XP_011530401.1 | 5 | 1089 | |||
XM_017008369.1 | XP_016863858.1 | 14 | 4349 | |||
XM_017008370.1 | XP_016863859.1 | 13 | 4129 | |||
XM_017008371.1 | XP_016863860.1 | 12 | 4009 | |||
XM_017008372.1 | XP_016863861.1 | 12 | 3388 | |||
XM_017008373.1 | XP_016863862.1 | 11 | 3325 | |||
GenBank mRNA | AK126054.1 | - | 4 | 738 | ||
BX647594.1 | - | 4 | 837 |