Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242636.1 | NP_001229565.1 | 12 | 1743 | ||
NM_001242637.1 | NP_001229566.1 | 12 | 1752 | |||
NM_001242638.1 | NP_001229567.1 | 14 | 2116 | |||
NM_001242639.1 | NP_001229568.1 | 13 | 1887 | |||
NM_139017.5 | NP_620586.3 | 12 | 1752 | |||
XM_011543142.2 | XP_011541444.1 | 12 | 1585 | |||
XM_011543143.2 | XP_011541445.1 | 12 | 1775 | |||
XM_011543144.2 | XP_011541446.1 | 11 | 1657 | |||
XM_011543145.2 | XP_011541447.1 | 9 | 1226 | |||
GenBank mRNA | AF106913.1 | - | 12 | 1601 | ||
AF486620.1 | - | 11 | 1464 | |||
AY358117.1 | - | 11 | 1464 | |||
AY499339.1 | - | 12 | 1625 | |||
AY499341.1 | - | 12 | 1625 | |||
AY499342.1 | - | 14 | 1999 | |||
BC110490.2 | - | 13 | 1679 | 40007489 |