Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145153.1 | NP_001138625.1 | 9 | 1487 | ||
NM_144649.2 | NP_653250.2 | 10 | 1619 | |||
GenBank mRNA | AK057631.1 | - | 10 | 1533 | ||
BC022053.2 | - | 6 | 1135 | |||
BC062592.1 | - | 10 | 1514 | 5224669 | ||
BC105783.1 | - | 9 | 1405 | 4669222 |