Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144723.2 | NP_653324.1 | 5 | 430 | ||
GenBank mRNA | AK055683.1 | - | 5 | 343 | ||
BC056668.1 | - | 5 | 347 | 4773709 | ||
BP210985.1 | - | 5 | 424 | |||
BP370587.1 | - | 4 | 286 | |||
X97303.1 | - | 2 | 70 |