Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145266.4 | NP_660309.1 | 4 | 543 | ||
GenBank mRNA | AK311957.1 | - | 4 | 506 | ||
BC017934.1 | - | 4 | 543 | 4698187 | ||
BX538290.1 | - | 4 | 544 | |||
FJ215684.1 | - | 4 | 483 |