Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166599.2 | NP_001160071.1 | 9 | 1992 | ||
NM_001170756.1 | NP_001164227.1 | 10 | 1249 | |||
NM_145284.5 | NP_660327.2 | 9 | 1195 | |||
XM_006724730.2 | XP_006724793.1 | 10 | 2108 | |||
XM_011531287.1 | XP_011529589.1 | 10 | 2120 | |||
XM_011531289.1 | XP_011529591.1 | 10 | 2117 | |||
GenBank mRNA | AK125991.1 | - | 10 | 1152 | ||
AK292517.1 | - | 9 | 1114 | |||
AL832218.1 | - | 9 | 1125 | |||
BC019221.2 | - | 9 | 1971 | 3841054 | ||
BC110846.1 | - | 9 | 2028 | 6046376 | ||
BX538218.1 | - | 10 | 1472 |