Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147189.2 | NP_671722.1 | 5 | 2055 | ||
XM_005251324.2 | XP_005251381.1 | 4 | 2049 | |||
XM_005251325.3 | XP_005251382.1 | 3 | 12212 | |||
XM_005251326.2 | XP_005251383.1 | 3 | 12237 | |||
XM_017013948.1 | XP_016869437.1 | 4 | 4624 | |||
GenBank mRNA | AK092221.1 | - | 5 | 2055 | ||
BC024294.1 | - | 2 | 1567 | 5440447 | ||
U79298.1 | - | 1 | 1186 |