Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317899.1 | NP_001304828.1 | 3 | 428 | ||
NM_152272.4 | NP_689485.1 | 4 | 1142 | |||
XM_006716415.2 | XP_006716478.1 | 3 | 598 | |||
XM_017013961.1 | XP_016869450.1 | 3 | 598 | |||
XM_017013962.1 | XP_016869451.1 | 5 | 1279 | |||
XM_017013963.1 | XP_016869452.1 | 3 | 405 | |||
XM_017013964.1 | XP_016869453.1 | 4 | 2291 | |||
XM_017013965.1 | XP_016869454.1 | 3 | 405 | |||
GenBank mRNA | AK021776.1 | - | 3 | 572 | ||
AK092500.1 | - | 3 | 378 | |||
AK097202.1 | - | 4 | 1139 | |||
AK296592.1 | - | 2 | 352 | |||
AK315664.1 | - | 3 | 579 | |||
AL832313.1 | - | 1 | 584 | |||
AL833843.1 | - | 3 | 547 | |||
BC004344.1 | - | 3 | 370 | |||
BC019110.1 | - | 3 | 544 | 5093453 | ||
BC042050.1 | - | 4 | 1123 | 5551762 |