Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317899.1 | NP_001304828.1 | 10 | 2574 | ||
NM_152272.4 | NP_689485.1 | 11 | 3288 | |||
XM_006716415.2 | XP_006716478.1 | 9 | 2683 | |||
XM_017013961.1 | XP_016869450.1 | 9 | 2958 | |||
XM_017013962.1 | XP_016869451.1 | 11 | 3639 | |||
XM_017013963.1 | XP_016869452.1 | 9 | 2765 | |||
XM_017013964.1 | XP_016869453.1 | 11 | 4437 | |||
XM_017013965.1 | XP_016869454.1 | 9 | 2490 | |||
GenBank mRNA | AK021776.1 | - | 10 | 2717 | ||
AK092500.1 | - | 10 | 2524 | |||
AK097202.1 | - | 11 | 3279 | |||
AL832313.1 | - | 7 | 2669 | |||
AL833843.1 | - | 10 | 2688 | |||
BC004344.1 | - | 10 | 2516 | |||
BC019110.1 | - | 10 | 2690 | 5093453 | ||
BC042050.1 | - | 11 | 3269 | 5551762 |