Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099271.1 | NP_001092741.1 | 12 | 1890 | ||
NM_152408.2 | NP_689621.2 | 11 | 1689 | |||
XM_005248436.1 | XP_005248493.1 | 12 | 1806 | |||
XM_011543158.2 | XP_011541460.1 | 11 | 1713 | |||
XM_011543159.1 | XP_011541461.1 | 13 | 1896 | |||
XM_011543160.1 | XP_011541462.1 | 11 | 1750 | |||
XM_017009037.1 | XP_016864526.1 | 11 | 1785 | |||
XM_017009038.1 | XP_016864527.1 | 11 | 1810 | |||
XM_017009039.1 | XP_016864528.1 | 11 | 1729 | |||
XM_017009040.1 | XP_016864529.1 | 10 | 1588 | |||
GenBank mRNA | AK093098.1 | - | 11 | 1659 | ||
AK296071.1 | - | 12 | 1807 | |||
BC101325.1 | - | 11 | 1655 | 40024984 | ||
BC101326.2 | - | 11 | 1659 | 40024987 | ||
BC101327.2 | - | 11 | 1659 | 40024989 | ||
BC101328.2 | - | 10 | 1479 |