Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286657.1 | NP_001273586.1 | 7 | 1445 | ||
NM_001286661.1 | NP_001273590.1 | 5 | 1005 | |||
NM_152417.2 | NP_689630.1 | 6 | 1383 | |||
XM_005251150.1 | XP_005251207.1 | 8 | 1573 | |||
XM_011517454.1 | XP_011515756.1 | 8 | 1597 | |||
XM_017013040.1 | XP_016868529.1 | 7 | 1157 | |||
GenBank mRNA | AK056932.1 | - | 6 | 1302 | ||
AL832935.1 | - | 6 | 1054 | |||
BC043417.1 | - | 4 | 3545 |