Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152548.2 | NP_689761.2 | 5 | 603 | ||
XM_005271906.4 | XP_005271963.2 | 6 | 626 | |||
XM_005271907.3 | XP_005271964.1 | 5 | 588 | |||
XM_011543207.1 | XP_011541509.1 | 5 | 598 | |||
XM_011543208.1 | XP_011541510.1 | 5 | 598 | |||
XM_011543209.1 | XP_011541511.1 | 3 | 284 | |||
XM_017009118.1 | XP_016864607.1 | 6 | 623 | |||
GenBank mRNA | AK058062.1 | - | 5 | 586 | ||
AK310245.1 | - | 4 | 405 | |||
BC034772.1 | - | 5 | 603 | 5169904 | ||
BM560335.1 | - | 4 | 408 |