Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256891.1 | NP_001243820.1 | 3, 4 | 262 | ||
NM_001256892.1 | NP_001243821.1 | 3, 4 | 238 | |||
NM_003130.3 | NP_003121.1 | 3, 4 | 262 | |||
NM_198901.1 | NP_944490.1 | 3, 4 | 238 | |||
XM_011516528.1 | XP_011514830.1 | 2, 3 | 132 | |||
GenBank mRNA | AK129770.1 | - | 3, 4 | 231 | ||
AK295225.1 | - | 3, 4 | 215 | |||
AK296601.1 | - | 3, 4 | 221 | |||
AK310366.1 | - | 3, 4 | 238 | |||
AL039198.1 | - | 3, 4 | 256 | |||
BC011025.1 | - | 3, 4 | 220 | 4281626 | ||
BI524104.1 | - | 3, 4 | 222 | |||
BI559592.1 | - | 3, 4 | 256 | |||
BI597920.1 | - | 3, 4 | 258 | |||
BM739144.1 | - | 3, 4 | 238 | |||
DA268050.1 | - | 3, 4 | 262 | |||
L12387.1 | - | 3, 4 | 208 | |||
M32886.1 | - | 3, 4 | 216 |