Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152577.3 | NP_689790.1 | 1 | 767 | ||
GenBank mRNA | AK098601.1 | - | 1 | 739 | ||
BC074910.2 | - | 1 | 709 | 30915257 | ||
BC126190.1 | - | 1 | 709 | 8991906 | ||
BC126192.1 | - | 1 | 709 | 8991908 | ||
GQ355336.1 | - | 1 | 767 |