Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136257.1 | NP_001129729.1 | 2 | 652 | ||
NM_001136258.1 | NP_001129730.1 | 3 | 844 | |||
NM_152621.5 | NP_689834.1 | 2 | 928 | |||
XM_011531698.2 | XP_011530000.1 | 3 | 982 | |||
XM_011531699.2 | XP_011530001.1 | 4 | 856 | |||
XM_011531700.2 | XP_011530002.1 | 3 | 753 | |||
XM_011531701.2 | XP_011530003.1 | 4 | 1867 | |||
XM_011531702.2 | XP_011530004.1 | 3 | 1795 | |||
XM_017007839.1 | XP_016863328.1 | 3 | 986 | |||
XM_017007840.1 | XP_016863329.1 | 4 | 1108 | |||
XM_017007841.1 | XP_016863330.1 | 3 | 982 | |||
GenBank mRNA | AF452717.1 | - | 3 | 747 | ||
AK290344.1 | - | 3 | 846 | |||
AK314049.1 | - | 2 | 643 | |||
BC028705.1 | - | 2 | 653 | 4823252 | ||
BC041369.2 | - | 2 | 921 | 5273650 | ||
BX649142.1 | - | 1 | 309 |