Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136257.1 | NP_001129729.1 | 5 | 1181 | ||
NM_001136258.1 | NP_001129730.1 | 6 | 1373 | |||
NM_152621.5 | NP_689834.1 | 5 | 1457 | |||
XM_011531698.2 | XP_011530000.1 | 6 | 1511 | |||
XM_011531699.2 | XP_011530001.1 | 7 | 1385 | |||
XM_011531700.2 | XP_011530002.1 | 6 | 1282 | |||
XM_011531701.2 | XP_011530003.1 | 7 | 2396 | |||
XM_011531702.2 | XP_011530004.1 | 6 | 2324 | |||
XM_017007839.1 | XP_016863328.1 | 6 | 1515 | |||
XM_017007840.1 | XP_016863329.1 | 7 | 1637 | |||
GenBank mRNA | AF452717.1 | - | 6 | 1276 | ||
AK290344.1 | - | 6 | 1375 | |||
AK314049.1 | - | 5 | 1172 | |||
AL833599.1 | - | 4 | 427 | |||
BC028705.1 | - | 5 | 1182 | 4823252 | ||
BC041369.2 | - | 5 | 1450 | 5273650 | ||
BX649142.1 | - | 4 | 838 |