Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152622.4 | NP_689835.3 | 9, 10 | 855 | ||
XM_011543217.1 | XP_011541519.1 | 9, 10 | 890 | |||
GenBank mRNA | BC041348.2 | - | 9, 10 | 852 | 5272287 | |
BX647359.1 | - | 9, 10 | 849 | |||
BX648294.1 | - | 9, 10 | 850 |