Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242377.1 | NP_001229306.1 | 7 | 939 | ||
NM_152624.5 | NP_689837.2 | 7 | 939 | |||
NR_038352.1 | 5 | 659 | ||||
XM_017009163.1 | XP_016864652.1 | 8 | 1031 | |||
GenBank mRNA | AK090564.1 | - | 7 | 854 | ||
AK296361.1 | - | 5 | 589 | |||
AY135173.1 | - | 7 | 913 | |||
AY146650.1 | - | 7 | 913 | |||
BC045596.1 | - | 3 | 236 | |||
BC064593.1 | - | 7 | 845 | 5757231 | ||
BG720562.1 | - | 5 | 666 | |||
BX648838.1 | - | 4 | 351 |