Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152628.3 | NP_689841.3 | 8, 9 | 840 | ||
XM_005250815.3 | XP_005250872.1 | 5, 6 | 479 | |||
XM_011516899.1 | XP_011515201.1 | 7, 8 | 817 | |||
XM_017013153.1 | XP_016868642.1 | 8, 9 | 840 | |||
XM_017013154.1 | XP_016868643.1 | 8, 9 | 840 | |||
XM_017013155.1 | XP_016868644.1 | 8, 9 | 840 | |||
XM_017013156.1 | XP_016868645.1 | 8, 9 | 840 | |||
XM_017013157.1 | XP_016868646.1 | 8, 9 | 840 | |||
XM_017013158.1 | XP_016868647.1 | 8, 9 | 840 | |||
XM_017013159.1 | XP_016868648.1 | 8, 9 | 840 | |||
XM_017013160.1 | XP_016868649.1 | 8, 9 | 840 | |||
XM_017013162.1 | XP_016868651.1 | 8, 9 | 840 | |||
XM_017013163.1 | XP_016868652.1 | 5, 6 | 920 | |||
GenBank mRNA | AK093906.1 | - | 8, 9 | 840 | ||
BC031260.1 | - | 5, 6 | 431 | 5272635 | ||
CD106024.1 | - | 3, 4 | 251 |