Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152628.3 | NP_689841.3 | 14 | 2152 | ||
XM_005250815.3 | XP_005250872.1 | 11 | 1791 | |||
XM_011516899.1 | XP_011515201.1 | 13 | 2129 | |||
XM_017013153.1 | XP_016868642.1 | 14 | 2149 | |||
XM_017013154.1 | XP_016868643.1 | 14 | 2149 | |||
XM_017013155.1 | XP_016868644.1 | 14 | 2146 | |||
XM_017013156.1 | XP_016868645.1 | 13 | 2095 | |||
XM_017013157.1 | XP_016868646.1 | 13 | 2038 | |||
XM_017013158.1 | XP_016868647.1 | 13 | 2035 | |||
XM_017013159.1 | XP_016868648.1 | 12 | 1981 | |||
XM_017013160.1 | XP_016868649.1 | 13 | 1948 | |||
XM_017013161.1 | XP_016868650.1 | 11 | 1903 | |||
XM_017013162.1 | XP_016868651.1 | 12 | 1834 | |||
XM_017013163.1 | XP_016868652.1 | 11 | 2232 | |||
GenBank mRNA | AK093906.1 | - | 14 | 2152 |