Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 2 | 410 | ||
NM_152632.3 | NP_689845.2 | 2 | 410 | |||
XM_017029452.1 | XP_016884941.1 | 2 | 403 | |||
XM_017029453.1 | XP_016884942.1 | 2 | 403 | |||
GenBank mRNA | AK126295.1 | - | 2 | 376 | ||
BC027936.1 | - | 2 | 363 | 5202137 | ||
CD686506.1 | - | 2 | 447 | |||
DA315191.1 | - | 2 | 403 |