Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003139.3 | NP_003130.2 | 2 | 336 | ||
XM_017018179.1 | XP_016873668.1 | 2 | 282 | |||
GenBank mRNA | AK312377.1 | - | 2 | 254 | ||
BC001162.1 | - | 2 | 216 | 3528505 | ||
BC008077.2 | - | 2 | 215 | |||
BC009110.1 | - | 2 | 179 | 3453825 | ||
BC013583.1 | - | 2 | 242 | 3877676 | ||
CR457025.1 | - | 2 | 158 |