Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152680.2 | NP_689893.1 | 7 | 2808 | ||
GenBank mRNA | AK056590.1 | - | 7 | 2682 | ||
BC019600.1 | - | 6 | 2411 |