Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256510.1 | NP_001243439.1 | 4 | 591 | ||
NM_001256511.1 | NP_001243440.1 | 4 | 472 | |||
NM_001256512.1 | NP_001243441.1 | 4 | 454 | |||
NM_001256513.1 | NP_001243442.1 | 4 | 382 | |||
NM_003143.2 | NP_003134.1 | 4 | 289 | |||
NR_046269.1 | 5 | 339 | ||||
XM_005250048.4 | XP_005250105.1 | 4 | 301 | |||
XM_005250049.4 | XP_005250106.1 | 4 | 340 | |||
XM_005250050.4 | XP_005250107.1 | 4 | 391 | |||
XM_005250051.4 | XP_005250108.1 | 4 | 417 | |||
GenBank mRNA | AF277319.1 | - | 4 | 235 | ||
AK294393.1 | - | 4 | 245 | |||
AK313033.1 | - | 4 | 302 | |||
BC000895.1 | - | 4 | 591 | 3464282 | ||
BC093054.1 | - | 4 | 251 | 30722138 | ||
BF131408.1 | - | 5 | 269 | |||
BG030060.1 | - | 405 | ||||
BP308841.1 | - | 3 | 158 | |||
BQ959702.1 | - | 4 | 354 | |||
BU532904.1 | - | 4 | 292 | |||
DB464012.1 | - | 4 | 378 | |||
M94556.1 | - | 4 | 234 |