Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277813.1 | NP_001264742.1 | 4 | 587 | ||
NM_001277814.1 | NP_001264743.1 | 4 | 341 | |||
NM_001277815.1 | NP_001264744.1 | 4 | 472 | |||
NM_001277817.1 | NP_001264746.1 | 3 | 280 | |||
NM_001277818.1 | NP_001264747.1 | 3 | 201 | |||
NM_152758.5 | NP_689971.4 | 3 | 441 | |||
NR_102434.1 | 3 | 441 | ||||
XM_011517509.2 | XP_011515811.1 | 4 | 434 | |||
GenBank mRNA | AK056219.1 | - | 3 | 351 | ||
AK093081.1 | - | 3 | 201 | |||
AK127574.1 | - | 3 | 373 | |||
AK298544.1 | - | 4 | 322 | |||
AL831816.1 | - | 3 | 276 | |||
AL832005.1 | - | 3 | 374 | |||
AL832335.1 | - | 4 | 370 | |||
AL832369.1 | - | 1 | 75 | |||
BC052970.1 | - | 3 | 390 | 6044054 | ||
BP223018.1 | - | 3 | 441 | |||
BP354748.1 | - | 3 | 281 | |||
BX537996.2 | - | 3 | 378 | |||
CN335163.1 | - | 3 | 428 | |||
DC419051.1 | - | 4 | 519 |