Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277813.1 | NP_001264742.1 | 5 | 725 | ||
NM_001277814.1 | NP_001264743.1 | 5 | 479 | |||
NM_001277815.1 | NP_001264744.1 | 5 | 610 | |||
NM_001277816.1 | NP_001264745.1 | 3 | 493 | |||
NM_001277817.1 | NP_001264746.1 | 4 | 418 | |||
NM_001277818.1 | NP_001264747.1 | 4 | 339 | |||
NM_152758.5 | NP_689971.4 | 4 | 579 | |||
XM_011517509.2 | XP_011515811.1 | 5 | 572 | |||
XM_011517510.2 | XP_011515812.1 | 3 | 335 | |||
GenBank mRNA | AK056219.1 | - | 4 | 489 | ||
AK093081.1 | - | 4 | 339 | |||
AK127574.1 | - | 4 | 511 | |||
AK298544.1 | - | 5 | 460 | |||
AL831816.1 | - | 4 | 414 | |||
AL832005.1 | - | 4 | 512 | |||
AL832335.1 | - | 5 | 508 | |||
AL832369.1 | - | 2 | 213 | |||
BC052970.1 | - | 4 | 528 | 6044054 | ||
BP354748.1 | - | 4 | 419 | |||
BX537996.2 | - | 4 | 516 |