Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277813.1 | NP_001264742.1 | 6 | 3784 | ||
NM_001277814.1 | NP_001264743.1 | 6 | 3538 | |||
NM_001277815.1 | NP_001264744.1 | 6 | 3669 | |||
NM_001277816.1 | NP_001264745.1 | 4 | 3552 | |||
NM_001277817.1 | NP_001264746.1 | 5 | 3477 | |||
NM_001277818.1 | NP_001264747.1 | 5 | 3398 | |||
NM_152758.5 | NP_689971.4 | 5 | 3638 | |||
NR_102434.1 | 4 | 2039 | ||||
XM_011517509.2 | XP_011515811.1 | 6 | 3631 | |||
XM_011517510.2 | XP_011515812.1 | 4 | 3394 | |||
GenBank mRNA | AK002173.1 | - | 1 | 315 | ||
AK127574.1 | - | 5 | 3570 | |||
AL831816.1 | - | 5 | 3473 | |||
AL832005.1 | - | 5 | 3571 | |||
AL832335.1 | - | 6 | 3566 | |||
AL832369.1 | - | 3 | 3272 | |||
BC052970.1 | - | 5 | 3587 | 6044054 | ||
BX537996.2 | - | 5 | 3574 |