Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152778.2 | NP_689991.1 | 3 | 261 | ||
XM_005262893.1 | XP_005262950.1 | 2 | 175 | |||
XM_005262896.1 | XP_005262953.1 | 2 | 175 | |||
XM_005262897.2 | XP_005262954.1 | 2 | 175 | |||
XM_005262898.2 | XP_005262955.1 | 2 | 175 | |||
XM_011531830.1 | XP_011530132.1 | 2 | 175 | |||
XM_011531831.1 | XP_011530133.1 | 2 | 175 | |||
XM_011531832.1 | XP_011530134.1 | 2 | 175 | |||
XM_017007989.1 | XP_016863478.1 | 2 | 175 | |||
GenBank mRNA | AK074564.1 | - | 2 | 110 | ||
AK294184.1 | - | 2 | 78 | |||
AK294439.1 | - | 2 | 107 | |||
AK294527.1 | - | 2 | 84 | |||
AK294957.1 | - | 2 | 103 | |||
AK315596.1 | - | 2 | 107 | |||
BC029503.1 | - | 3 | 226 | 5272664 | ||
BI553701.1 | - | 2 | 176 |