Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168465.1 | NP_001161937.1 | 17 | 2810 | ||
NM_001168466.1 | NP_001161938.1 | 15 | 2552 | |||
NM_001168467.1 | NP_001161939.1 | 16 | 2524 | |||
NM_152780.3 | NP_689993.2 | 16 | 2687 | |||
XM_005274478.2 | XP_005274535.1 | 17 | 2673 | |||
XM_005274479.2 | XP_005274536.1 | 17 | 2640 | |||
XM_005274480.3 | XP_005274537.1 | 16 | 2574 | |||
XM_006724481.2 | XP_006724544.1 | 18 | 2820 | |||
XM_006724482.2 | XP_006724545.1 | 17 | 2697 | |||
XM_011545483.1 | XP_011543785.1 | 18 | 2787 | |||
XM_011545484.1 | XP_011543786.1 | 18 | 2770 | |||
XM_011545485.1 | XP_011543787.1 | 18 | 2705 | |||
XM_017029393.1 | XP_016884882.1 | 17 | 2664 | |||
XM_017029394.1 | XP_016884883.1 | 17 | 2647 | |||
XM_017029395.1 | XP_016884884.1 | 17 | 2623 | |||
XM_017029396.1 | XP_016884885.1 | 17 | 2582 | |||
XM_017029397.1 | XP_016884886.1 | 17 | 2558 | |||
GenBank mRNA | AK027409.1 | - | 16 | 2589 | ||
AK098768.1 | - | 10 | 1243 | |||
AK127056.1 | - | 14 | 2226 | |||
AK316117.1 | - | 15 | 2460 | |||
BC037165.2 | - | 14 | 2354 | |||
BC089400.1 | - | 17 | 2731 | 3063898 | ||
BC136379.1 | - | 16 | 2604 | 9020366 |