Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040057.2 | NP_001035146.1 | 12 | 1665 | ||
NM_001288584.1 | NP_001275513.1 | 11 | 1650 | |||
NM_152789.3 | NP_690002.2 | 11 | 1566 | |||
NR_109929.1 | 11 | 1525 | ||||
XM_011516004.2 | XP_011514306.1 | 11 | 1829 | |||
XM_011516005.2 | XP_011514307.1 | 11 | 3150 | |||
GenBank mRNA | AK125498.1 | - | 9 | 1946 | ||
AL833918.2 | - | 12 | 1665 | |||
BC032461.1 | - | 11 | 1511 | 5197918 | ||
BC057771.1 | - | 12 | 1656 | 5301001 | ||
BC090868.1 | - | 11 | 1550 | 6190536 |
Gene Symbol: FAM133CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027508.1 | 1 | 1996 |
Gene Symbol: FAM133DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034169.1 | 1 | 1510 |