Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 883 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 994 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 907 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 1205 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 886 | |||
NM_153345.3 | NP_699176.1 | 3 | 785 | |||
NR_040003.2 | 2 | 943 | ||||
NR_104250.1 | 3 | 624 | ||||
NR_104251.1 | 2 | 523 | ||||
NR_104252.1 | 3 | 621 | ||||
NR_104253.1 | 2 | 547 | ||||
NR_104254.1 | 3 | 645 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 730 | ||
AK172816.1 | - | 5 | 936 | |||
AY358781.1 | - | 3 | 800 | |||
BC016719.1 | - | 1 | 155 | |||
BC035517.1 | - | 3 | 1185 | 5207904 | ||
BC035888.1 | - | 2 | 546 | |||
BE562464.1 | - | 2 | 530 | |||
BF036500.1 | - | 1 | 443 | |||
BI761527.1 | - | 2 | 473 | |||
CB156055.1 | - | 2 | 463 | |||
DT220400.1 | - | 2 | 475 |