Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 1515 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 1626 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 1539 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 1837 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 1518 | |||
NM_153345.3 | NP_699176.1 | 3 | 1417 | |||
NR_040003.2 | 2 | 1575 | ||||
NR_104250.1 | 3 | 1256 | ||||
NR_104251.1 | 2 | 1155 | ||||
NR_104252.1 | 3 | 1253 | ||||
NR_104253.1 | 2 | 1179 | ||||
NR_104254.1 | 3 | 1277 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 1362 | ||
AK172816.1 | - | 5 | 1569 | |||
BC035517.1 | - | 3 | 1817 | 5207904 | ||
BC035888.1 | - | 2 | 1178 |