Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 888 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 999 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 912 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 1210 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 891 | |||
NM_153345.3 | NP_699176.1 | 3 | 790 | |||
NR_040003.2 | 2 | 948 | ||||
NR_104250.1 | 3 | 629 | ||||
NR_104251.1 | 2 | 528 | ||||
NR_104252.1 | 3 | 626 | ||||
NR_104253.1 | 2 | 552 | ||||
NR_104254.1 | 3 | 650 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 735 | ||
AK172816.1 | - | 5 | 941 | |||
AY358781.1 | - | 3 | 805 | |||
BC016719.1 | - | 1 | 160 | |||
BC035517.1 | - | 3 | 1190 | 5207904 | ||
BC035888.1 | - | 2 | 551 | |||
BE562464.1 | - | 2 | 535 | |||
BF036500.1 | - | 1 | 448 | |||
BI761527.1 | - | 2 | 478 | |||
CB156055.1 | - | 2 | 468 |