Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM161B SIRNA Assays ›
Gene Symbol:
TMEM161B
Gene Name:
transmembrane protein 161B
Gene Aliases:
FLB3342, PRO1313
Chromosome Location:
Chr.5: 88189419 - 88268917 on Build GRCh38
UniGene ID:
Hs.379972
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128184
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM161B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289007.1
NP_001275936.1
7
806
NM_001289008.1
NP_001275937.1
8
938
NM_153354.4
NP_699185.1
7
806
NR_110266.1
8
893
XM_006714553.1
XP_006714616.1
7
756
XM_006714554.3
XP_006714617.1
6
693
XM_006714555.3
XP_006714618.1
5
575
XM_006714556.3
XP_006714619.2
5
536
XM_011543201.2
XP_011541503.1
6
673
XM_011543202.2
XP_011541504.2
5
1218
XM_011543203.2
XP_011541505.2
3
1023
XM_011543204.2
XP_011541506.2
5
491
XM_017009093.1
XP_016864582.1
7
756
XM_017009094.1
XP_016864583.1
6
647
XM_017009095.1
XP_016864584.1
7
756
XM_017009096.1
XP_016864585.1
6
677
XM_017009097.1
XP_016864586.1
6
680
XM_017009098.1
XP_016864587.1
6
625
XM_017009099.1
XP_016864588.1
5
554
XM_017009100.1
XP_016864589.1
6
647
XM_017009101.1
XP_016864590.1
6
609
XM_017009102.1
XP_016864591.1
6
610
XM_017009103.1
XP_016864592.1
6
677
XM_017009104.1
XP_016864593.1
6
680
XM_017009105.1
XP_016864594.1
7
718
XM_017009106.1
XP_016864595.1
6
610
XM_017009107.1
XP_016864596.1
6
640
XM_017009108.1
XP_016864597.1
5
525
XM_017009109.1
XP_016864598.1
6
690
XM_017009110.1
XP_016864599.1
6
676
GenBank mRNA
AK299896.1
-
7
708
AK300006.1
-
3
235
AK300893.1
-
8
888
AK301091.1
-
7
757
AY358693.1
-
7
764
BC037287.2
-
7
775
4837154
BM479365.1
-
4
415
BX647345.1
-
8
813
BX648757.1
-
6
706
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008770407, XM_008760644.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline