Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM161B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289008.1 | NP_001275937.1 | 13 | 1779 | ||
NM_153354.4 | NP_699185.1 | 12 | 1647 | |||
NR_110266.1 | 13 | 1747 | ||||
XM_006714553.1 | XP_006714616.1 | 11 | 1500 | |||
XM_006714554.3 | XP_006714617.1 | 11 | 1534 | |||
XM_006714555.3 | XP_006714618.1 | 10 | 1416 | |||
XM_011543201.2 | XP_011541503.1 | 11 | 1514 | |||
XM_017009103.1 | XP_016864592.1 | 11 | 1518 | |||
XM_017009105.1 | XP_016864594.1 | 12 | 1559 | |||
XM_017009106.1 | XP_016864595.1 | 11 | 1451 | |||
XM_017009107.1 | XP_016864596.1 | 11 | 1481 | |||
XM_017009108.1 | XP_016864597.1 | 10 | 1366 | |||
XM_017009109.1 | XP_016864598.1 | 10 | 1434 | |||
XM_017009110.1 | XP_016864599.1 | 10 | 1420 | |||
GenBank mRNA | AK300006.1 | - | 8 | 995 | ||
AK300893.1 | - | 13 | 1729 | |||
BC037287.2 | - | 12 | 1616 | 4837154 | ||
BX647345.1 | - | 13 | 1667 | |||
BX648757.1 | - | 9 | 6630 |